Containers, square, with screw cap
Supplier: DELTALAB
Certificates
About this item
Containers and insert plugs made of HDPE, caps are made of PP.
- Good chemical and temperature resistance
- Tagging points on neck and cap for a tamper proof closure or for labelling
- All models have moulded-in graduations
Translucent square containers with star-shaped caps.
Product Information
- Catalog #Supplier #CapacityW×D×HColourNeck-ØPkAvailabilityPrice
- DELT44460844460825 ml32×34×52 mmTranslucent white18 mm500Log in for availability1 * 500 KS16 220,00 KčSupplier Article Number: 444608Local Article Number: DELT444608Specifications:
Specifications
Environmentally Preferable - DELT44460944460950 ml38×38×70 mmTranslucent white23 mm400Log in for availability1 * 400 KS11 340,00 KčSupplier Article Number: 444609Local Article Number: DELT444609Specifications:
Specifications
Environmentally Preferable - Supplier Article Number: 444610Local Article Number: DELT444610Specifications:
Specifications
Environmentally Preferable - DELT444611444611250 ml57×60×110 mmTranslucent white33 mm1Log in for availability1 * 1 KS88,00 KčSupplier Article Number: 444611Local Article Number: DELT444611Specifications:
Specifications
Environmentally Preferable - DELT444612444612500 ml70×80×138 mmTranslucent white44 mm1Log in for availability1 * 1 KS88,00 KčSupplier Article Number: 444612Local Article Number: DELT444612Specifications:
Specifications
Environmentally Preferable - DELT4446134446131000 ml80×103×176 mmTranslucent white58 mm1Log in for availability1 * 1 KS128,00 KčSupplier Article Number: 444613Local Article Number: DELT444613Specifications:
Specifications
Environmentally Preferable - DELT4446144446142000 ml100×134×210 mmTranslucent white58 mm40Log in for availability1 * 40 KS8 380,00 KčSupplier Article Number: 444614Local Article Number: DELT444614Specifications: