Tisk…

5-(5-Fluoro-2-hydroxyphenyl)isoxazole, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL543683-1GEA 2720 CZK
SIAL543683-1G
5-(5-Fluoro-2-hydroxyphenyl)isoxazole, Sigma-Aldrich®
5-(5-Fluoro-2-hydroxyphenyl)isoxazole
Vzorec: C₉H₆FNO₂
MDL Number: MFCD03070563
CAS číslo: 288401-62-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR