Tisk…

4-Fluoro-2-methylbenzenesulphonamide, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL559652-1GEA 0 CZK
SIAL559652-1G
4-Fluoro-2-methylbenzenesulphonamide, Sigma-Aldrich®
4-Fluoro-2-methylbenzenesulphonamide
Vzorec: C₇H₈FNO₂S
MDL Number: MFCD02709831
CAS číslo: 489-17-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR