Tisk…

4-Chloroquinazoline, Sigma-Aldrich®

Dodavatel: Merck

SIAL700096-1GEA 1460 CZK
SIAL700096-1G
4-Chloroquinazoline, Sigma-Aldrich®
4-Chloroquinazoline
Vzorec: C₈H₅ClN₂
MW: 164,5900 g/mol
MDL Number: MFCD00228682
CAS číslo: 5190-68-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR