5,7-Dichloro-2-methyl-8-quinolinol
Dodavatel: EHRENSTORFER GMBH
EHERC11505800EA
2890
CZK
EHERC11505800
5,7-Dichloro-2-methyl-8-quinolinol
5,7-Dichloro-2-methyl-8-quinolinol
Vzorec:
C₁₀H₇Cl₂NO MW: 228,0747 g/mol |
MDL Number:
MFCD00023984 CAS číslo: 72-80-0 EINECS: 200-789-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...