Tisk…
(Trimethylphosphine)gold(I) chloride, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
SIAL404217-1GEA 0 CZK
SIAL404217-1G SIAL404217-250MG
(Trimethylphosphine)gold(I) chloride, Sigma-Aldrich®
(Trimethylphosphine)gold(I) chloride
Vzorec: C₃H₉AuClP
MW: 308,5 g/mol
MDL Number: MFCD00058861
CAS číslo: 15278-97-4