Tisk…

5-Chloro-2-(3-chloromethyl-4H-1,2,4-triazol-4-yl)benzophenone

Dodavatel: U.S. Pharmacopeia

USPH1249916EA 35650 CZK
USPH1249916
5-Chloro-2-(3-chloromethyl-4H-1,2,4-triazol-4-yl)benzophenone
5-Chloro-2-(3-chloromethyl-4H-1,2,4-triazol-4-yl)benzophenone
Vzorec: C₁₆H₁₁Cl₂N₃O
CAS číslo: 37945-06-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR