Tisk…

Dimetridazole-[1-methyl-D3], Supelco®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

34196-10MGEA 0 CZK
34196-10MG
Dimetridazole-[1-methyl-D3], Supelco®
Dimetridazole-[1-methyl-D3]
Vzorec: C₅H₄D₃N₃O₂
MDL Number: MFCD09265282
CAS číslo: 64678-69-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR