Tisk…

Tin(II) fluorophosphate

Dodavatel: Thermo Scientific
Nebezpečí

39608.14EA 6080 CZK
39608.14 39608.06
Tin(II) fluorophosphate
Tin(II) fluorophosphate
Vzorec: FO₃PSn
Storage Temperature: Ambient
MDL Number: MFCD00058794
CAS číslo: 52262-58-5
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR