Tisk…

4-(4-Methyl-1-piperazinylcarbonyl)phenylboronic acid pinacol ester, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL706175-5GEA 0 CZK
SIAL706175-5G SIAL706175-1G
4-(4-Methyl-1-piperazinylcarbonyl)phenylboronic acid pinacol ester, Sigma-Aldrich®
4-(4-Methyl-1-piperazinylcarbonyl)phenylboronic acid pinacol ester
Vzorec: C₁₈H₂₇BN₂O₃
MW: 330,23
MDL Number: MFCD05863914
CAS číslo: 832114-06-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR