Tisk…

1-(3-Chlorophenyl)biguanide hydrochloride, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

SIALC144-500MGEA 0 CZK
SIALC144-500MG
1-(3-Chlorophenyl)biguanide hydrochloride, Sigma-Aldrich®
1-(3-Chlorophenyl)biguanide hydrochloride
Vzorec: C₈H₁₁Cl₂N₅
MW: 248,12 g/mol
MDL Number: MFCD00053019
CAS číslo: 2113-05-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR