2,2'-Biquinoline analytical reagent, Supelco®
M-Clarity™ quality level = MQ100
1.02955.0005EA
0
CZK
1.02955.0005
2,2'-Biquinoline analytical reagent, Supelco®
2,2'-Biquinoline
Vzorec:
C₁₈H₁₂N₂ MW: 256,31 g/mol |
MDL Number:
MFCD00006740 CAS číslo: 119-91-5 EINECS: 204-357-5 |
Výsledky specifikačního testu
Solubility | 1.02 mg/l (25 °C) |
Melting Point | 192 - 195 °C |
Molar Mass | 256.3 g/mol |
Bulk Density | 290 kg/m3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...