Tisk…

Lithium tantalum(V) isopropoxide ≥99.9% (metals basis)

Dodavatel: Thermo Scientific
Nebezpečí

40520.09EA 13940 CZK
40520.09 40520.04 40520.18
Lithium tantalum(V) isopropoxide ≥99.9% (metals basis)
Lithium tantalum(V) isopropoxide
Vzorec: C₁₈H₄₂LiO₆Ta
Bod varu: 160…180 °C (0,1 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00798538
CAS číslo: 21864-24-4
UN: 2925
ADR: 4.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR