Tisk…

2-(Tributylstannyl)benzo[b]thiophene, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí


M-Clarity™ quality level = MQ100

SIAL683930-1GEA 5820 CZK
SIAL683930-1G
2-(Tributylstannyl)benzo[b]thiophene, Sigma-Aldrich®
2-(Tributylstannyl)benzo[b]thiophene
Vzorec: C₂₀H₃₂SSn
MW: 423,22 g/mol
MDL Number: MFCD01318936
CAS číslo: 148961-88-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR