Tisk…

(R)-(-)-3-Aminopiperidine dihydrochloride, Sigma-Aldrich®

Dodavatel: Merck

SIAL15626-1G-FEA 0 CZK
SIAL15626-1G-F
(R)-(-)-3-Aminopiperidine dihydrochloride, Sigma-Aldrich®
(R)-(-)-3-Aminopiperidine dihydrochloride
Vzorec: C₅H₁₄Cl₂N₂
MW: 173,08 g/mol
MDL Number: MFCD06799458
CAS číslo: 334618-23-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR