Tisk…

1-(Cyanoacetyl)piperidine, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL102180-100GEA 12870 CZK
SIAL102180-100G
1-(Cyanoacetyl)piperidine, Sigma-Aldrich®
1-(Cyanoacetyl)piperidine
Vzorec: C₈H₁₂N₂O
MW: 152,20 g/mol
MDL Number: MFCD00006487
CAS číslo: 15029-30-8
EINECS: 239-111-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR