Tisk…

1-Boc-piperidine-4-carboxylic acid, Sigma-Aldrich®

Dodavatel: Merck

SIAL15518-5GEA 0 CZK
SIAL15518-5G
1-Boc-piperidine-4-carboxylic acid, Sigma-Aldrich®
1-Boc-piperidine-4-carboxylic acid
Vzorec: C₁₁H₁₉NO₄
MW: 229,3 g/mol
MDL Number: MFCD00076999
CAS číslo: 84358-13-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR