3-(2-Aminoethylamino)propylmethyldimethoxysilane, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
SIAL06667-25MLEA
1050
CZK
SIAL06667-25ML
3-(2-Aminoethylamino)propylmethyldimethoxysilane, Sigma-Aldrich®
3-(2-Aminoethylamino)propylmethyldimethoxysilane
Vzorec:
C₈H₂₂N₂O₂Si MW: 206,36 g/mol |
MDL Number:
MFCD00054776 CAS číslo: 3069-29-2 EINECS: 221-336-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...