(3-Aminopropyl)dimethylethoxysilane, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
SIAL588857-1GEA
3300
CZK
SIAL588857-1G
SIAL588857-5G
(3-Aminopropyl)dimethylethoxysilane, Sigma-Aldrich®
(3-Aminopropyl)dimethylethoxysilane
Vzorec:
C₇H₁₉NOSi MW: 161,3174 g/mol |
MDL Number:
MFCD00053947 CAS číslo: 18306-79-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...