Tisk…

N(α)-Boc-D-tryptophan, Sigma-Aldrich®

Dodavatel: Merck

SIAL15185-5GEA 2410 CZK
SIAL15185-5G
N(α)-Boc-D-tryptophan, Sigma-Aldrich®
N(α)-Boc-D-tryptophan
Vzorec: C₁₆H₂₀N₂O₄
MW: 304,3 g/mol
MDL Number: MFCD00037944
CAS číslo: 5241-64-5
EINECS: 226-042-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR