N(α)-Boc-L-tryptophan ≥99.0% (by TLC), Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Boc-Trp-OH
SIAL15512-5G-FEA
878
CZK
SIAL15512-5G-F
N(α)-Boc-L-tryptophan ≥99.0% (by TLC), Sigma-Aldrich®
N(α)-Boc-L-tryptophan
Vzorec:
C₁₆H₂₀N₂O₄ MW: 304,3 g/mol |
MDL Number:
MFCD00065595 CAS číslo: 13139-14-5 EINECS: 236-072-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...