1-Methyl-1H-pyrazol-4-carbaldehyde, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
SIAL654043-1GEA
0
CZK
SIAL654043-1G
1-Methyl-1H-pyrazol-4-carbaldehyde, Sigma-Aldrich®
1-Methyl-1H-pyrazol-4-carbaldehyde
Vzorec:
C₅H₆N₂O MW: 110,12 g/mol |
MDL Number:
MFCD00460465 CAS číslo: 25016-11-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...