Tisk…

4-Methyl-2-(methylthio)pyrimidine, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL674885-10GEA 3430 CZK
SIAL674885-10G
4-Methyl-2-(methylthio)pyrimidine, Sigma-Aldrich®
4-Methyl-2-(methylthio)pyrimidine
Vzorec: C₆H₈N₂S
MW: 140,21 g/mol
MDL Number: MFCD00023242
CAS číslo: 14001-63-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR