Sodium aurothiomalate ≥99.9% (metals basis)
Dodavatel: Thermo Scientific
|
Varování
|
39740.03EA
12800
CZK
39740.03
Sodium aurothiomalate ≥99.9% (metals basis)
Sodium aurothiomalate
Vzorec:
C₄H₃AuNa₂O₄S Storage Temperature: Ambient |
MDL Number:
MFCD00064304 CAS číslo: 12244-57-4 EINECS: 235-479-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...