Tisk…
Sodium aurothiomalate, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
SIAL157201-250MGEA 7250 CZK
SIAL157201-250MG SIAL157201-1G
Sodium aurothiomalate, Sigma-Aldrich®
Sodium aurothiomalate
Vzorec: C₄H₆AuNaO₄S
MW: 370,11 g/mol
MDL Number: MFCD00064304
CAS číslo: 12244-57-4
EINECS: 235-479-7