Tisk…

N-Boc-D-alaninol, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL469505-5GEA 0 CZK
SIAL469505-5G
N-Boc-D-alaninol, Sigma-Aldrich®
N-Boc-D-alaninol
Vzorec: C₈H₁₇NO₃
MW: 175,2 g/mol
MDL Number: MFCD00235912
CAS číslo: 106391-86-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR