N(α)-Benzyloxycarbonyl-L-lysine
MOLE11422112-5GEA
1790
CZK
MOLE11422112-5G
N(α)-Benzyloxycarbonyl-L-lysine
N(α)-Benzyloxycarbonyl-L-lysine
Vzorec:
C₁₄H₂₀N₂O₄ MW: 280,3 g/mol |
MDL Number:
MFCD00038204 CAS číslo: 2212-75-1 EINECS: 218-662-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...