Tisk…

(S)-3-(Boc-amino)-3-(3-methoxyphenyl)propionic acid, Sigma-Aldrich®

Dodavatel: Merck

SIAL55894-500MG-FEA 13310 CZK
SIAL55894-500MG-F
(S)-3-(Boc-amino)-3-(3-methoxyphenyl)propionic acid, Sigma-Aldrich®
(S)-3-(Boc-amino)-3-(3-methoxyphenyl)propionic acid
Vzorec: C₁₅H₂₁NO₅
MW: 295,3309 g/mol
MDL Number: MFCD03427899
CAS číslo: 499995-77-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR