(S)-3-(Boc-amino)-3-(2-(trifluoromethyl)phenyl)propionic acid, Sigma-Aldrich®
Dodavatel: Merck
SIAL55685-500MG-FEA
0
CZK
SIAL55685-500MG-F
(S)-3-(Boc-amino)-3-(2-(trifluoromethyl)phenyl)propionic acid, Sigma-Aldrich®
(S)-3-(Boc-amino)-3-(2-(trifluoromethyl)phenyl)propionic acid
Vzorec:
C₁₅H₁₈F₃NO₄ MW: 333,3029 g/mol |
MDL Number:
MFCD03427925 CAS číslo: 500770-77-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...