Tisk…

4-(Pentafluorothio)benzonitrile ≥97%

Dodavatel: Thermo Scientific
Nebezpečí

H35537.06EA 6830 CZK
H35537.06 H35537.03
4-(Pentafluorothio)benzonitrile ≥97%
4-(Pentafluorothio)benzonitrile
Vzorec: C₇H₄F₅NS
Storage Temperature: Ambient
MDL Number: MFCD03788521
CAS číslo: 401892-85-1
UN: 3439
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR