3-Sulphobenzoic acid disodium salt monohydrate ≥97%
Dodavatel: Thermo Scientific
B24154.22EA
1610
CZK
B24154.22
3-Sulphobenzoic acid disodium salt monohydrate ≥97%
3-Sulphobenzoic acid disodium salt monohydrate
Vzorec:
C₇H₄Na₂O₅S·1H₂O Storage Temperature: Ambient |
MDL Number:
MFCD00149379 CAS číslo: 14995-40-5 EINECS: 239-081-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...