4-Sulphobenzoic acid monopotassium salt, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
SIAL310638-50GEA
1690
CZK
SIAL310638-50G
4-Sulphobenzoic acid monopotassium salt, Sigma-Aldrich®
4-Sulphobenzoic acid monopotassium salt
Vzorec:
C₇H₅KO₅S MW: 240,28 g/mol |
MDL Number:
MFCD00007509 CAS číslo: 5399-63-3 EINECS: 226-435-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...