Tisk…

4-Methoxyazobenzene, Sigma-Aldrich®

Dodavatel: Merck

SIAL64760-5G-FEA 0 CZK
SIAL64760-5G-F
4-Methoxyazobenzene, Sigma-Aldrich®
4-Methoxyazobenzene
Vzorec: C₁₃H₁₂N₂O
MW: 212,2472 g/mol
MDL Number: MFCD00039804
CAS číslo: 2396-60-3
EINECS: 219-250-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR