Tisk…

4-Methoxy-N,N-diphenylaniline, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL646121-1GEA 0 CZK
SIAL646121-1G SIAL646121-10G
4-Methoxy-N,N-diphenylaniline, Sigma-Aldrich®
4-Methoxy-N,N-diphenylaniline
Vzorec: C₁₉H₁₇NO
MDL Number: MFCD03929020
CAS číslo: 4316-51-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR