Tisk…

1,6-Diphenoxy-2,4-hexadiyne, Sigma-Aldrich®

Dodavatel: Merck

SIAL42655-10GEA 0 CZK
SIAL42655-10G
1,6-Diphenoxy-2,4-hexadiyne, Sigma-Aldrich®
1,6-Diphenoxy-2,4-hexadiyne
Vzorec: C₁₈H₁₄O₂
MW: 262,31
MDL Number: MFCD00041712
CAS číslo: 30980-37-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR