Tisk…

6-Amino-1-methyluracil, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL346799-25GEA 2670 CZK
SIAL346799-25G
6-Amino-1-methyluracil, Sigma-Aldrich®
6-Amino-1-methyluracil
Vzorec: C₅H₇N₃O₂
MW: 141,13 g/mol
MDL Number: MFCD00075366
CAS číslo: 2434-53-9
EINECS: 219-422-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR