Tisk…

N-Cbz-L-phenylalaninol, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL421723-5GEA 0 CZK
SIAL421723-5G
N-Cbz-L-phenylalaninol, Sigma-Aldrich®
N-Cbz-L-phenylalaninol
Vzorec: C₁₇H₁₉NO₃
MW: 285,3 g/mol
MDL Number: MFCD00191138
CAS číslo: 6372-14-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR