Tisk…

(±)-Cbz-α-phosphonoglycine trimethyl ester, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

SIAL376353-5GEA 8070 CZK
SIAL376353-5G
(±)-Cbz-α-phosphonoglycine trimethyl ester, Sigma-Aldrich®
(±)-Cbz-α-phosphonoglycine trimethyl ester
Vzorec: C₁₃H₁₈NO₇P
MW: 331,27 g/mol
MDL Number: MFCD00043304
CAS číslo: 88568-95-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR