Tisk…

4,4'-Biphenyldiboronic acid, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL456799-500MGEA 0 CZK
SIAL456799-500MG
4,4'-Biphenyldiboronic acid, Sigma-Aldrich®
4,4'-Biphenyldiboronic acid
Vzorec: C₁₂H₁₂B₂O₄
MW: 241,85 g/mol
MDL Number: MFCD00151795
CAS číslo: 4151-80-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR