Calmagite
MOLE28767001-25GEA
4170
CZK
MOLE28767001-25G
Calmagite
Calmagite
Vzorec:
C₁₇H₁₄N₂O₅S MW: 358,37 g/mol |
MDL Number:
MFCD00011656 CAS číslo: 3147-14-6 EINECS: 221-563-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...