Tisk…

2-Acetyl-6-bromopyridine

Dodavatel: Apollo Scientific

APOSOR12051-5GEA 1170 CZK
APOSOR12051-5G
2-Acetyl-6-bromopyridine
2-Acetyl-6-bromopyridine
Vzorec: C₇H₆BrNO
MW: 200,03
MDL Number: MFCD00272200
CAS číslo: 49669-13-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR