Tisk…

5-Nitro-3-picoline ≥97%

Dodavatel: Thermo Scientific
Nebezpečí

H31683.06EA 8550 CZK
H31683.06
5-Nitro-3-picoline ≥97%
5-Nitro-3-picoline
Vzorec: C₆H₆N₂O₂
MW: 138,13 g/mol
Bod tání: 94…98 °C
Storage Temperature: Ambient
MDL Number: MFCD04114214
CAS číslo: 6960-20-9
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR