Tisk…

4-(3-(Ethoxycarbonyl)piperidine-1-carbonyl)phenylboronic acid pinacol ester ≥98%

Dodavatel: Thermo Scientific

H28086.03EA 930 CZK
H28086.03 H28086.06
4-(3-(Ethoxycarbonyl)piperidine-1-carbonyl)phenylboronic acid pinacol ester ≥98%
4-(3-(Ethoxycarbonyl)piperidine-1-carbonyl)phenylboronic acid pinacol ester
Vzorec: C₂₁H₃₀BNO₅
MW: 387,28
Storage Temperature: Ambient
MDL Number: MFCD06657751
CAS číslo: 850411-14-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR