Tisk…

4-Fluoro-3-(1-pyrrolidinylcarbonyl)phenylboronic acid ≥98%

Nebezpečí

H52481.MDEA 3570 CZK
H52481.MD H52481.03
4-Fluoro-3-(1-pyrrolidinylcarbonyl)phenylboronic acid ≥98%
4-Fluoro-3-(1-pyrrolidinylcarbonyl)phenylboronic acid
Vzorec: C₁₁H₁₃BFNO₃
MW: 237,04 g/mol
Bod tání: 152…154 °C
Storage Temperature: Ambient
MDL Number: MFCD08235050
CAS číslo: 874219-31-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR