Tisk…

(R)-(-)-2-Hydroxy-2-phenylpropionic acid ≥98%

Dodavatel: Thermo Scientific

L08644.MDEA 1760 CZK
L08644.MD L08644.03
(R)-(-)-2-Hydroxy-2-phenylpropionic acid ≥98%
(R)-(-)-2-Hydroxy-2-phenylpropionic acid
Vzorec: C₉H₁₀O₃
Bod tání: 115…117 °C
Storage Temperature: Ambient
MDL Number: MFCD00067699
CAS číslo: 3966-30-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR