Tisk…

(R)-(+)-N,N-Dimethyl-1-phenylethanamine, Sigma-Aldrich®

Dodavatel: Merck

SIAL242071-1GEA 0 CZK
SIAL242071-1G
(R)-(+)-N,N-Dimethyl-1-phenylethanamine, Sigma-Aldrich®
(R)-(+)-N,N-Dimethyl-1-phenylethanamine
Vzorec: C₁₀H₁₅N
MDL Number: MFCD00008857
CAS číslo: 19342-01-9
EINECS: 242-976-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR