Tisk…

3-Iodo-4-methylbenzonitrile, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL642401-25GEA 7280 CZK
SIAL642401-25G
3-Iodo-4-methylbenzonitrile, Sigma-Aldrich®
3-Iodo-4-methylbenzonitrile
Vzorec: C₈H₆IN
MDL Number: MFCD06797817
CAS číslo: 42872-79-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR