5-Chlorobenzimidazole, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
SIAL631248-5GEA
3490
CZK
SIAL631248-5G
SIAL631248-1G
5-Chlorobenzimidazole, Sigma-Aldrich®
5-Chlorobenzimidazole
Vzorec:
C₇H₅ClN₂ MW: 152,5810 g/mol |
MDL Number:
MFCD17015012 CAS číslo: 4887-82-5 EINECS: 225-503-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...