Tisk…

4-Chlorodiphenyl sulphone, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL194115-100GEA 9660 CZK
SIAL194115-100G
4-Chlorodiphenyl sulphone, Sigma-Aldrich®
4-Chlorodiphenyl sulphone
Vzorec: C₁₂H₉ClO₂S
MDL Number: MFCD00007549
CAS číslo: 80-00-2
EINECS: 201-242-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR