Tisk…

4-Bromo-2,3,5,6-tetrafluorobenzonitrile, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL344052-5GEA 8680 CZK
SIAL344052-5G
4-Bromo-2,3,5,6-tetrafluorobenzonitrile, Sigma-Aldrich®
4-Bromo-2,3,5,6-tetrafluorobenzonitrile
Vzorec: C₇BrF₄N
MW: 253,9792 g/mol
MDL Number: MFCD00075282
CAS číslo: 17823-40-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR